Learn More
Chemplex Industries PreAssembled Sample Cup Trimless Sleeve, Vented Cap
Catalog No. NC1591783
Encompass
PreAssembled Sample Cup Trimless Sleeve, Vented Cap, 1.61" (40.8mm) O.D. X 1.51" (38.4mm) High, with 6.6um Mylar; 16 Sets/tube. Eliminates the need for the Oxford L242 aluminum holder.
Catalog No. NC1591783 Supplier Chemplex Industries Supplier No. 10P40OX14
PreAssembled Sample Cup Trimless Sleeve, Vented Cap, 1.61" (40.8mm) O.D. X 1.51" (38.4mm) High, with 6.6um Mylar; 16 Sets/tube. Eliminates the need for the Oxford L242 aluminum holder.