Learn More
TARGETMOL CHEMICALS INC XRK3F2 5MG
SDP
Catalog No. 502581358
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. XRK3F2 is an inhibitor of the p62-ZZ domain blunts MM-induced Runx2 suppression in vitro and induces new bone formation and remodeling in the presence of tumor in vivo. purity: 98%
Catalog No. 50-258-1358 Supplier TARGETMOL CHEMICALS INC Supplier No. T133605MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. XRK3F2 is an inhibitor of the p62-ZZ domain blunts MM-induced Runx2 suppression in vitro and induces new bone formation and remodeling in the presence of tumor in vivo. purity: 98%