Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XRP44X, Tocris Bioscience™
Description
Ras-Net (Elk-3) pathway inhibitor (IC50 = 10 - 20 nM). Indirectly inhibits Net phosphorylation upstream of Erk1/2 activation. Mediates G2-M cell cycle arrest, suppresses growth of multiple cell types (IC50 ∽ 2 nM), inhibits angiogenesis and acts as a microtubule depolymerizing agent in vitro.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 380.87 |
| Percent Purity | >99% |
| Chemical Name or Material | Ras GTPase Inhibitors |
Safety and Handling
| Recommended Storage | Store at Room Temperature |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?