Learn More
Aviva Systems Biology Corp XRRA1 Antibody - N-terminal region (ARP53441_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53441_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against XRRA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human XRRA1. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-63033