Learn More
TWD Scientific LLC Extra Small Antistatic Polypropylene Weighing Funnel. Case of 1152.
SDP
Catalog No. 501173376 Shop All TWD Scientific LLC Products
Encompass_Preferred
Extra small disposable antistatic polypropylene weighing funnel. Fits 96-well microtubes. Funnels have a maximum capacity of 1mL Solid/0.5mL Liquid 1152/Case
Catalog No. 50-117-3376 Supplier TWD Scientific LLC Supplier No. ASWFXSCS
Extra small disposable antistatic polypropylene weighing funnel. Fits 96-well microtubes. Funnels have a maximum capacity of 1mL Solid/0.5mL Liquid 1152/Case