Learn More
TWD Scientific LLC Extra Small Antistatic Polypropylene Weighing Funnel. Pack of 96.
SDP
Catalog No. 501173375 Shop All TWD Scientific LLC Products
Encompass_Preferred
Extra small disposable antistatic polypropylene weighing funnel. Fits 96-well microtubes. Funnels have a maximum capacity of 1mL Solid/0.5mL Liquid. 96/Pack
Catalog No. 50-117-3375 Supplier TWD Scientific LLC Supplier No. ASWFXSPK
Extra small disposable antistatic polypropylene weighing funnel. Fits 96-well microtubes. Funnels have a maximum capacity of 1mL Solid/0.5mL Liquid. 96/Pack