Learn More
TWD Scientific LLC Extra Small Polypropylene Weighing Funnels. Case of 1152
SDP
Catalog No. 501173370 Shop All TWD Scientific LLC Products
Encompass_Preferred
Disposable polypropylene weighing funnel, size extra small. Fits 96-well microtubes.Funnels have a maximum capacity of 1mL Solid/0.5mL Liquid. 1152/Case
Catalog No. 50-117-3370 Supplier TWD Scientific LLC Supplier No. DPWFPPXSCS
Disposable polypropylene weighing funnel, size extra small. Fits 96-well microtubes.Funnels have a maximum capacity of 1mL Solid/0.5mL Liquid. 1152/Case