Learn More
TWD Scientific LLC Extra Small Glass Weighing Funnel. Pack of 6
SDP
Catalog No. 501173385 Shop All TWD Scientific LLC Products
Encompass_Preferred
Disposable borosilicate glass weighing funnel, size extra small. Fits 96-well microtubes. Funnels have a maximum capacity of 1mL Solid/0.5mL Liquid. 6/Pack
Catalog No. 50-117-3385 Supplier TWD Scientific LLC Supplier No. GWFBSXSPK
Disposable borosilicate glass weighing funnel, size extra small. Fits 96-well microtubes. Funnels have a maximum capacity of 1mL Solid/0.5mL Liquid. 6/Pack