Learn More
Cole-Parmer Glass Weighing Funnels, micro, 6/pk (Glass)
Catalog No. NC0044204 Shop All Cole-Parmer Products
Encompass_Preferred
These reusable borosilicate glass weighing funnels are designed to ensure complete sample transfer to volumetric flasks. Funnels are flat on one side so you can set them on a balance. Micro funnel is sized to fit microwell plates.
Catalog No. NC0044204 Supplier Cole-Parmer Supplier No. 3470000
These reusable borosilicate glass weighing funnels are designed to ensure complete sample transfer to volumetric flasks. Funnels are flat on one side so you can set them on a balance. Micro funnel is sized to fit microwell plates.