Learn More
Waters Corp Waters XSelect Column, CSH C18, 2.5 μm, 4.6 mm × 150 mm
Supplier:  Waters Corp 186007038
Encompass_Preferred
These CSH130 C18 based particles provide excellent peak shape for peptides with FA- or TFA-containing mobile phases and show little dependence on strong ion pairing agents, making them ideal for LC or LC-MS applications. Packed in ultra-low dispersion hardware, these 2.5 μm eXtended Performance columns ensure exceptional separation performance and throughput for HPLC assays, supporting a seamless transition to UPLC adoption.
  • Charged Surface Hybrid (CSH) technology offers superior peak shape and increased loading capacity for basic compounds.
  • Allows for higher resolution separations of complex samples.
  • Fully compatible with HPLC, UPLC, and UHPLC technologies.
  • Withstands pressures up to 10,000 psi (690 Bar).
  • Offers outstanding peak capacities for peptides and small proteins up to approximately 10,000 Daltons.
Catalog No. 50-596-964