Learn More
Waters Corp Waters Corporation XSelect CSH C18 Column, 130Å, 3.5 μm
Supplier:  Waters Corp 186005644
Encompass_Preferred
XSelect CSH C18 columns are general-purpose columns offering alternative selectivity compared to other C18 sorbents. They utilize Charged Surface Hybrid (CSH) technology to address issues such as peak shape asymmetry, poor loading for basic compounds, column bleed, and slow mobile-phase re-equilibration.
  • Can be used as general purpose columns that provide alternative selectivity compared to other C18 sorbents.
  • Charged Surface Hybrid (CSH) technology helps overcome column-related problems such as peak shape asymmetry and poor loading for basic compounds, column bleed, and slow mobile-phase re-equilibration.
Catalog No. 50-466-174