Learn More
Waters Corp XSelect CSH C18 Column, 130Å, 3.5 μm, 3 mm X 75 mm
Catalog No. 50466177 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect CSH C18 columns can be used as general purpose columns that provide alternative selectivity compared to other C18 sorbents. Charged Surface Hybrid (CSH) technology helps overcome column related problems such as peak shape asymmetry and poor loading for basic compounds, column bleed, and slow mobile-phase re-equilibration.
  • Can be used as general purpose columns.
  • Provide alternative selectivity compared to other C18 sorbents.
  • Charged Surface Hybrid (CSH) technology helps overcome column related problems.
  • Addresses peak shape asymmetry.
  • Addresses poor loading for basic compounds.
  • Addresses column bleed.
  • Addresses slow mobile-phase re-equilibration.
Catalog No. 50-466-177 Supplier Waters Corp Supplier No. 186005647
XSelect CSH C18 columns can be used as general purpose columns that provide alternative selectivity compared to other C18 sorbents. Charged Surface Hybrid (CSH) technology helps overcome column related problems such as peak shape asymmetry and poor loading for basic compounds, column bleed, and slow mobile-phase re-equilibration.
  • Can be used as general purpose columns.
  • Provide alternative selectivity compared to other C18 sorbents.
  • Charged Surface Hybrid (CSH) technology helps overcome column related problems.
  • Addresses peak shape asymmetry.
  • Addresses poor loading for basic compounds.
  • Addresses column bleed.
  • Addresses slow mobile-phase re-equilibration.