Learn More
Waters Corp Waters Column, CSH C18, 130Å, 2.5 μm, 2.1 mm × 150 mm
Supplier:  Waters Corp 186006727
Encompass_Preferred
This CSH C18 Column is designed for high-performance liquid chromatography (HPLC) assays, offering exceptional separation performance and robustness. Featuring Charged Surface Hybrid (CSH) technology, it provides superior peak shape and increased loading capacity for basic compounds, and is engineered to withstand 18000 psi (1240 Bar).
  • Alternative selectivity compared to other C18 sorbents.
  • Charged Surface Hybrid (CSH) technology for superior peak shape and increased loading capacity for basic compounds.
  • Ultra-low dispersion hardware packing.
  • 2.5 μm eXtended Performance [XP] for exceptional separation, robustness, and throughput in HPLC assays.
  • Enables seamless transition to UPLC adoption.
  • Engineered to endure 18000 psi (1240 Bar).
  • Exceptional performance and prolonged operational lifespan.
Catalog No. 50-466-435