Learn More
Waters Corp Waters CSH C18 Column, 130 Å, 2.5 μm, 2.1 x 150 mm
Supplier:  Waters Corp 176002891
Encompass_Preferred
XSelect CSH C18 columns are general-purpose C18 columns that offer alternative selectivity compared to other C18 sorbents. The Charged Surface Hybrid (CSH) technology provides superior peak shape and increased loading capacity for basic compounds. These 2.5 μm eXtended Performance [XP] columns offer exceptional separation performance, robustness, and throughput for HPLC assays.
  • Superior peak shape and increased loading capacity for basic compounds with Charged Surface Hybrid (CSH) technology.
  • Ultra-low dispersion hardware enables exceptional separation performance and robustness.
  • Designed to withstand pressures up to 18000 psi (1240 Bar).
  • Wide pH operating range of 1 to 11.
  • Low silanol activity and endcapping for enhanced column life.
  • Seamless transition path for UPLC adoption.
Catalog No. 50-465-931