Learn More
Waters Corp XSelect CSH C18 Method Validation Kit, 130Å, 2.5 µm, 2.1 mm X 150 mm, 3/pk
Catalog No. 50466477 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit consists of three XSelect CSH C18 columns packed with three different batches of sorbent. XSelect CSH C18 columns are general purpose C18 columns that provide alternative selectivity compared to other C18 sorbents. Charged Surface Hybrid (CSH) technology enables superior peak shape and increased loading capacity for basic compounds. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
Catalog No. 50-466-477 Supplier Waters Corp Supplier No. 186006785
This method validation kit consists of three XSelect CSH C18 columns packed with three different batches of sorbent. XSelect CSH C18 columns are general purpose C18 columns that provide alternative selectivity compared to other C18 sorbents. Charged Surface Hybrid (CSH) technology enables superior peak shape and increased loading capacity for basic compounds. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.