Learn More
Waters Corp HPLC Column, XSelect CSH C18, 2.5 μm, 3.0 x 150 mm
Catalog No. 50466478 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters XSelect CSH C18 HPLC column is engineered for high-performance liquid chromatography applications. Featuring Charged Surface Hybrid (CSH) technology, this column provides excellent peak shape for basic compounds and enhanced loading capacity. Its 2.5 μm particle size ensures high efficiency and resolution for demanding separations.
  • Engineered for high-performance liquid chromatography
  • Utilizes Charged Surface Hybrid (CSH) technology
  • Provides excellent peak shape for basic compounds
  • Ensures enhanced loading capacity
  • Delivers high efficiency and resolution
  • Suitable for demanding separations
Catalog No. 50-466-478 Supplier Waters Corp Supplier No. 186006786
Add to Cart
Add to Cart
The Waters XSelect CSH C18 HPLC column is engineered for high-performance liquid chromatography applications. Featuring Charged Surface Hybrid (CSH) technology, this column provides excellent peak shape for basic compounds and enhanced loading capacity. Its 2.5 μm particle size ensures high efficiency and resolution for demanding separations.
  • Engineered for high-performance liquid chromatography
  • Utilizes Charged Surface Hybrid (CSH) technology
  • Provides excellent peak shape for basic compounds
  • Ensures enhanced loading capacity
  • Delivers high efficiency and resolution
  • Suitable for demanding separations