Learn More
Waters Corp Waters Corporation XSelect CSH C18 Method Validation Kit, 130 Å, 2.5 μm, 4.6 mm x 150 mm
Catalog No. 50466479 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The method validation kit includes three XSelect CSH C18 columns, each packed with a different sorbent batch. These are general-purpose C18 columns offering alternative selectivity compared to other C18 sorbents. The Charged Surface Hybrid (CSH) technology incorporated provides superior peak shape and increased loading capacity for basic compounds.
  • Includes three columns, each packed with a different batch of sorbent.
  • General-purpose C18 columns offer alternative selectivity.
  • Charged Surface Hybrid (CSH) technology provides superior peak shape and increased loading capacity for basic compounds.
  • Packed in ultra-low dispersion hardware, the 2.5 μm eXtended Performance [XP] columns deliver exceptional separation performance, robustness, and throughput for HPLC assays.
  • Enables a seamless transition path toward UPLC adoption.
Catalog No. 50-466-479 Supplier Waters Corp Supplier No. 186006787
Add to Cart
Add to Cart
The method validation kit includes three XSelect CSH C18 columns, each packed with a different sorbent batch. These are general-purpose C18 columns offering alternative selectivity compared to other C18 sorbents. The Charged Surface Hybrid (CSH) technology incorporated provides superior peak shape and increased loading capacity for basic compounds.
  • Includes three columns, each packed with a different batch of sorbent.
  • General-purpose C18 columns offer alternative selectivity.
  • Charged Surface Hybrid (CSH) technology provides superior peak shape and increased loading capacity for basic compounds.
  • Packed in ultra-low dispersion hardware, the 2.5 μm eXtended Performance [XP] columns deliver exceptional separation performance, robustness, and throughput for HPLC assays.
  • Enables a seamless transition path toward UPLC adoption.