Learn More
Waters Corp HPLC Column, XSelect CSH C18, 3 mm x 150 mm, 2.5 μm
Supplier: Waters Corp 186006728
XSelect CSH C18 columns are general purpose C18 columns offering alternative selectivity. Charged Surface Hybrid (CSH) technology ensures excellent peak shape and increased loading capacity for basic compounds. These 2.5 μm eXtended Performance (XP) columns are designed for exceptional separation performance, robustness, and high throughput in HPLC assays, with seamless UPLC transition.
- Provides alternative C18 selectivity.
- CSH technology for excellent peak shape and increased loading capacity for basic compounds.
- Ultra-low dispersion hardware delivers exceptional separation performance and high throughput.
- Supports seamless transition to UPLC.
- Inner diameter of 3 mm and length of 150 mm.
- Particle size of 2.5 μm.
- Designed for reproducible, highly selective separations with CSH and HSS particle technologies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.