Learn More
Waters Corp Waters CSH C18 Column, 130Å, 2.5 μm, 4.6 mm, 150 mm
Supplier:  Waters Corp 186006729
Encompass_Preferred
These CSH C18 columns are general-purpose C18 columns offering alternative selectivity and superior peak shape for basic compounds via Charged Surface Hybrid (CSH) technology. They are packed in ultra-low dispersion hardware for exceptional HPLC performance and facilitate seamless UPLC adoption.
  • General purpose C18 columns with alternative selectivity.
  • Charged Surface Hybrid (CSH) technology for superior peak shape and increased loading capacity.
  • Ultra-low dispersion hardware for exceptional separation performance and throughput.
  • Seamless transition to UPLC adoption.
  • Compatible with HPLC, UHPLC, and UPLC pressures.
  • Manufactured in cGMP ISO, 9001 certified facilities ensuring reproducibility.
Catalog No. 50-466-437