Learn More
Waters Corp WATERS CORPORATION C18 Column, 130Å, 3.5μm, 4.6 mm x 150 mm
Supplier:  Waters Corp 186005270
Encompass_Preferred
XSelect CSH C18 columns are general-purpose columns that offer alternative selectivity compared to other C18 sorbents. They incorporate Charged Surface Hybrid (CSH) technology to address common column-related issues such as peak shape asymmetry, poor loading for basic compounds, column bleed, and slow mobile-phase re-equilibration. These columns are designed for a wide selectivity range and offer seamless transferability across different particle sizes and column formats.
  • Provides alternative selectivity compared to other C18 sorbents.
  • Charged Surface Hybrid (CSH) technology helps overcome peak shape asymmetry and poor loading for basic compounds.
  • Offers superior peak shape for basic compounds.
  • Enhanced loading capacity.
  • Improved batch-to-batch reproducibility.
Catalog No. NC0005679