Learn More
Waters Corp XSelect CSH C18 Column, 3.5 μm, 130 Å, 3 mm x 150 mm, eConnect
Supplier:  Waters Corp 186005263RF
Encompass_Preferred
XSelect CSH C18 columns offer alternative selectivity to other C18 sorbents. They use Charged Surface Hybrid (CSH) technology to address peak shape asymmetry, poor loading for basic compounds, column bleed, and slow mobile-phase re-equilibration. This column includes an eConnect tag for automatic identification, tracking, and digital tracing. Full advantages are realized with the Waters Alliance iS System and Empower software.
  • Alternative selectivity to other C18 sorbents.
  • CSH technology resolves peak shape asymmetry and poor loading for basic compounds.
  • Reduces column bleed; improves mobile-phase re-equilibration.
  • eConnect tag for automatic identification, tracking, and digital tracing.
  • Optimized with specific systems and software.
Catalog No. 50-239-2650