Learn More
Waters Corp Waters Column, C18, 130Å, 5 μm, 4.6 mm × 150 mm
Catalog No. 502037695 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect CSH C18 columns offer alternative selectivity using Charged Surface Hybrid (CSH) technology. CSH addresses peak shape asymmetry, poor loading for basic compounds, and slow mobile-phase re-equilibration. It provides superior peak shape, high peak capacity, and rapid column equilibration, enabling high-resolution separations and excellent batch-to-batch reproducibility for HPLC assays.
  • Alternative selectivity compared to other C18 sorbents.
  • Overcomes peak shape asymmetry and poor loading for basic compounds.
  • Superior peak shape and high peak capacity.
  • Increased mass load for preparative separation without ion-pairing reagents.
  • Rapid column equilibration.
  • High-resolution separations for complex samples.
  • Excellent batch-to-batch reproducibility.
  • Operational pH range: 1 to 11.
  • Withstands pressure of 6000 psi (415 Bar).
Catalog No. 50-203-7695 Supplier Waters Corp Supplier No. 186005290
XSelect CSH C18 columns offer alternative selectivity using Charged Surface Hybrid (CSH) technology. CSH addresses peak shape asymmetry, poor loading for basic compounds, and slow mobile-phase re-equilibration. It provides superior peak shape, high peak capacity, and rapid column equilibration, enabling high-resolution separations and excellent batch-to-batch reproducibility for HPLC assays.
  • Alternative selectivity compared to other C18 sorbents.
  • Overcomes peak shape asymmetry and poor loading for basic compounds.
  • Superior peak shape and high peak capacity.
  • Increased mass load for preparative separation without ion-pairing reagents.
  • Rapid column equilibration.
  • High-resolution separations for complex samples.
  • Excellent batch-to-batch reproducibility.
  • Operational pH range: 1 to 11.
  • Withstands pressure of 6000 psi (415 Bar).