Learn More
Waters Corp Waters XSelect CSH C18 Column, 130Å, 3.5 μm, 1 mm × 100 mm
Supplier:  Waters Corp 186005250
Encompass_Preferred
XSelect CSH C18 columns serve as general-purpose columns, offering alternative selectivity compared to other C18 sorbents. The Charged Surface Hybrid (CSH) technology incorporated in these columns helps address common column-related issues such as peak shape asymmetry, poor loading for basic compounds, column bleed, and slow mobile-phase re-equilibration.
  • General purpose columns that offer alternative selectivity compared to other C18 sorbents.
  • Charged Surface Hybrid (CSH) technology addresses common column-related issues such as peak shape asymmetry and poor loading for basic compounds, column bleed, and slow mobile-phase re-equilibration.
Catalog No. 50-791-289