Learn More
Waters Corp XSelect CSH C18 Column, 130Å, 3.5 μm, 2.1 mm x 100 mm
Catalog No. 50791295 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These general-purpose CSH C18 columns provide alternative selectivity for reversed-phase separations. Featuring Charged Surface Hybrid (CSH) technology, they resolve common chromatographic issues and deliver high-resolution separation for complex samples. Compatible with HPLC, UPLC, and UHPLC, these columns offer excellent batch-to-batch consistency and enhanced loading capacity.
  • Offers alternative selectivity compared to other C18 sorbents.
  • CSH technology improves peak shape, loading, and re-equilibration.
  • Achieves higher resolution separations for complex samples.
  • Compatible with HPLC, UPLC, and UHPLC technologies.
  • Ensures excellent batch-to-batch consistency.
  • Provides better loading capacity and particle size scalability.
  • Ultra-low dispersion hardware supports UPLC adoption.
Catalog No. 50-791-295 Supplier Waters Corp Supplier No. 186005256
These general-purpose CSH C18 columns provide alternative selectivity for reversed-phase separations. Featuring Charged Surface Hybrid (CSH) technology, they resolve common chromatographic issues and deliver high-resolution separation for complex samples. Compatible with HPLC, UPLC, and UHPLC, these columns offer excellent batch-to-batch consistency and enhanced loading capacity.
  • Offers alternative selectivity compared to other C18 sorbents.
  • CSH technology improves peak shape, loading, and re-equilibration.
  • Achieves higher resolution separations for complex samples.
  • Compatible with HPLC, UPLC, and UHPLC technologies.
  • Ensures excellent batch-to-batch consistency.
  • Provides better loading capacity and particle size scalability.
  • Ultra-low dispersion hardware supports UPLC adoption.