Learn More
Waters Corp XSelect CSH C18 Column, 130Å, 3.5 µm, 2.1 mm X 150 mm, 1/pk
Catalog No. 50791296 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect CSH C18 columns can be used as general purpose columns that provide alternative selectivity compared to other C18 sorbents. Charged Surface Hybrid (CSH) technology helps you overcome column related problems such as peak shape asymmetry and poor loading for basic compounds, column bleed, and slow mobile-phase re-equilibration.
Catalog No. 50-791-296 Supplier Waters Corp Supplier No. 186005257
XSelect CSH C18 columns can be used as general purpose columns that provide alternative selectivity compared to other C18 sorbents. Charged Surface Hybrid (CSH) technology helps you overcome column related problems such as peak shape asymmetry and poor loading for basic compounds, column bleed, and slow mobile-phase re-equilibration.