Learn More
Waters Corp Waters Column, C18, 3.5 μm, 2.1 mm × 50 mm, 130 Å
Catalog No. 50791294 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This C18 column offers alternative selectivity, using CSH technology for improved peak shape, loading, reduced column bleed, and faster re-equilibration.
  • Provides diverse analyte selectivity and high-resolution separations for complex samples.
  • Exceptional separation performance, robustness, and throughput across HPLC, UPLC, and UHPLC.
  • Superior peak shape for basic compounds, low column bleed, and ultra-efficiencies.
  • Excellent batch-to-batch reproducibility for rapid method development.
  • Outstanding performance, equivalent backpressure to core-shell columns, improved loading capacity and scalability.
  • CSH technology enhances LC performance in acidic, low ionic strength mobile phases; non-end-capped sorbent enhances acidic compound retention.
  • Ultra-low dispersion hardware ensures outstanding separation performance, supporting UPLC adoption.
Catalog No. 50-791-294 Supplier Waters Corp Supplier No. 186005255
This C18 column offers alternative selectivity, using CSH technology for improved peak shape, loading, reduced column bleed, and faster re-equilibration.
  • Provides diverse analyte selectivity and high-resolution separations for complex samples.
  • Exceptional separation performance, robustness, and throughput across HPLC, UPLC, and UHPLC.
  • Superior peak shape for basic compounds, low column bleed, and ultra-efficiencies.
  • Excellent batch-to-batch reproducibility for rapid method development.
  • Outstanding performance, equivalent backpressure to core-shell columns, improved loading capacity and scalability.
  • CSH technology enhances LC performance in acidic, low ionic strength mobile phases; non-end-capped sorbent enhances acidic compound retention.
  • Ultra-low dispersion hardware ensures outstanding separation performance, supporting UPLC adoption.