Learn More
Waters Corp Waters XSelect CSH C18 Column, 130 Å, 3.5 μm, 4.6 mm × 100 mm
Supplier:  Waters Corp 186005269
Encompass_Preferred
XSelect CSH C18 columns offer alternative selectivity using Charged Surface Hybrid (CSH) technology. They address issues like peak shape asymmetry and poor loading for basic compounds, enabling high-resolution separations for complex samples.
  • Charged Surface Hybrid (CSH) technology provides superior peak shape for basic compounds and high peak capacity.
  • Enables high-resolution separations with exceptional robustness, performance, and throughput for HPLC assays.
  • Offers excellent batch-to-batch reproducibility, improved loading capacity, and particle size scalability.
  • Allows rapid column equilibration and quick changes in mobile-phase conditions and pH.
  • Operates within a pH range of 1 to 11.
  • Withstands pressures up to 6000 psi (415 Bar).
Catalog No. 50-791-307