Learn More
Waters Corp Waters Column, C18, 130Å, 5 μm, 3 × 30 mm
Catalog No. 50791315 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect CSH C18 columns are general purpose columns that offer alternative selectivity compared to other C18 sorbents. They incorporate Charged Surface Hybrid (CSH) technology, which helps to address common column-related issues such as peak shape asymmetry and poor loading for basic compounds, column bleed, and slow mobile-phase re-equilibration.
  • Provides alternative selectivity when compared to other C18 sorbents.
  • Employs Charged Surface Hybrid (CSH) technology to mitigate issues like peak shape asymmetry, poor loading for basic compounds, column bleed, and slow mobile-phase re-equilibration.
Catalog No. 50-791-315 Supplier Waters Corp Supplier No. 186005279
Add to Cart
Add to Cart
XSelect CSH C18 columns are general purpose columns that offer alternative selectivity compared to other C18 sorbents. They incorporate Charged Surface Hybrid (CSH) technology, which helps to address common column-related issues such as peak shape asymmetry and poor loading for basic compounds, column bleed, and slow mobile-phase re-equilibration.
  • Provides alternative selectivity when compared to other C18 sorbents.
  • Employs Charged Surface Hybrid (CSH) technology to mitigate issues like peak shape asymmetry, poor loading for basic compounds, column bleed, and slow mobile-phase re-equilibration.