Learn More
Waters Corp Waters Corporation Column, XSelect CSH C18, 5 μm, 4.6 mm x 100 mm, 130Å
Supplier:  Waters Corp 186005289
Encompass_Preferred
XSelect CSH C18 columns offer alternative selectivity to other C18 sorbents. Their Charged Surface Hybrid (CSH) technology addresses common column issues like peak shape asymmetry and slow mobile-phase re-equilibration. These columns deliver improved resolution for complex samples, are compatible with HPLC, UPLC, and UHPLC systems, and ensure good batch-to-batch consistency.
  • Offers unsurpassed selectivity and repeatability for chromatographic analysis.
  • Provides better loading capacity and particle size scalability due to CSH technology.
  • Includes ultra-low dispersion hardware, enabling outstanding separation performance and a smooth transition path toward UPLC adoption.
Catalog No. 50-791-324