Learn More
Waters Corp Waters Corporation Column, CSH C18, 3.0 x 50 mm, 5 μm
Supplier:  Waters Corp 186005280
Encompass_Preferred
XSelect CSH C18 columns leverage Charged Surface Hybrid technology to enhance LC column performance across various mobile phases. They are ideal for diverse chromatographic separations, offering robust and scalable solutions for analytical and preparative needs.
  • Charged Surface Hybrid (CSH) technology improves LC column performance in acidic, low ionic strength mobile phases.
  • Broad selectivity for a wide range of chromatographic separations.
  • High analyte loading and operational stability.
  • Full scalability for UPLC®, HPLC, and preparative platforms.
  • Highly pressure-tolerant hybrid materials for rugged performance and extended column lifetimes.
  • Improves peak shape and mass load when using formic acid mobile phases.
  • Seamless transferability across UPLC, analytical HPLC, and preparative HPLC.
Catalog No. 50-791-316