Learn More
Waters Corp Waters CSH C18 Column, 130Å, 5 µm, 4.6 mm × 30 mm
Supplier:  Waters Corp 186005286
Encompass_Preferred
XSelect CSH C18 columns can be used as general purpose columns that provide alternative selectivity compared to other C18 sorbents. Charged Surface Hybrid (CSH) technology helps overcome column-related problems such as peak shape asymmetry and poor loading for basic compounds, column bleed, and slow mobile-phase re-equilibration.
  • General purpose columns providing alternative selectivity compared to other C18 sorbents.
  • Incorporates Charged Surface Hybrid (CSH) technology.
  • Helps overcome peak shape asymmetry for basic compounds.
  • Addresses poor loading for basic compounds.
  • Reduces column bleed.
  • Improves mobile-phase re-equilibration.
Catalog No. 50-791-322