Learn More
Waters Corp Waters Sentry Guard Cartridge, 3.5 μm, 130 Å, 4.6 mm x 20 mm
Supplier:  Waters Corp 186005264
Encompass_Preferred
These CSH C18 Sentry Guard Cartridges are designed to protect analytical columns by shielding against particulate contamination.
Catalog No. 50-791-302