Learn More
Waters Corp Waters Column, XSelect CSH C18, 3.5 μm, 4.6 x 20 mm
Catalog No. 50791303 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect CSH C18 columns are general-purpose columns that provide alternative selectivity compared to other C18 sorbents. They incorporate Charged Surface Hybrid (CSH) technology, which helps to mitigate common column-related issues.
  • Can be used as general purpose columns, offering alternative selectivity compared to other C18 sorbents.
  • Utilizes Charged Surface Hybrid (CSH) technology.
  • Addresses issues such as peak shape asymmetry and poor loading for basic compounds.
  • Reduces column bleed and slow mobile-phase re-equilibration.
Catalog No. 50-791-303 Supplier Waters Corp Supplier No. 186005265
XSelect CSH C18 columns are general-purpose columns that provide alternative selectivity compared to other C18 sorbents. They incorporate Charged Surface Hybrid (CSH) technology, which helps to mitigate common column-related issues.
  • Can be used as general purpose columns, offering alternative selectivity compared to other C18 sorbents.
  • Utilizes Charged Surface Hybrid (CSH) technology.
  • Addresses issues such as peak shape asymmetry and poor loading for basic compounds.
  • Reduces column bleed and slow mobile-phase re-equilibration.