Learn More
Waters Corp Waters Method Validation Kit, XSelect CSH C18, 3 pk
Catalog No. 50791498 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit contains three high-performance columns, each with a different gel lot of stationary phase material, designed to ensure robust method development. Utilizing Charged Surface Hybrid (CSH) Technology, these columns provide high-resolution separations, improved peak shapes for basic compounds, and superior performance in acidic, low ionic strength, and LC/MS friendly mobile phases.
  • Contains three columns with different gel lots for method validation.
  • Employs Charged Surface Hybrid (CSH) Technology.
  • Delivers superior peak shape for basic compounds and high peak capacity.
  • Supports high-resolution separations of complex samples.
  • Offers robustness, performance, and high throughput for HPLC assays.
  • Achieves rapid column equilibration and increased preparative mass load.
  • Ensures excellent batch-to-batch reproducibility.
Catalog No. 50-791-498 Supplier Waters Corp Supplier No. 186005541
This method validation kit contains three high-performance columns, each with a different gel lot of stationary phase material, designed to ensure robust method development. Utilizing Charged Surface Hybrid (CSH) Technology, these columns provide high-resolution separations, improved peak shapes for basic compounds, and superior performance in acidic, low ionic strength, and LC/MS friendly mobile phases.
  • Contains three columns with different gel lots for method validation.
  • Employs Charged Surface Hybrid (CSH) Technology.
  • Delivers superior peak shape for basic compounds and high peak capacity.
  • Supports high-resolution separations of complex samples.
  • Offers robustness, performance, and high throughput for HPLC assays.
  • Achieves rapid column equilibration and increased preparative mass load.
  • Ensures excellent batch-to-batch reproducibility.