Learn More
Waters Corp Waters CSH C18 Method Validation Kit, 130Å, 5 μm, 2.1 mm x 150 mm
Catalog No. 50791500 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect HPLC columns, powered by CSH Technology, provide a broad range of selectivities and excellent performance in acidic, low ionic strength, and LC/MS friendly mobile phases. The Method Validation Kits include three columns, each containing a different gel lot of stationary phase material.
  • C18 chemistry for reversed phase separation
  • Hybrid particle substrate
  • pH range from 1 to 11
  • Maximum pressure of 6000 psi (415 bar)
  • Endcapped for improved performance
  • Low silanol activity
  • Spherical particle shape
  • 5 μm particle size
  • 130 Å pore size
  • Method validation kit format
  • Designed for HPLC systems with CSH particle technology
  • 2.1 mm inner diameter and 150 mm length
  • Units per package: 3
Catalog No. 50-791-500 Supplier Waters Corp Supplier No. 186005543
XSelect HPLC columns, powered by CSH Technology, provide a broad range of selectivities and excellent performance in acidic, low ionic strength, and LC/MS friendly mobile phases. The Method Validation Kits include three columns, each containing a different gel lot of stationary phase material.
  • C18 chemistry for reversed phase separation
  • Hybrid particle substrate
  • pH range from 1 to 11
  • Maximum pressure of 6000 psi (415 bar)
  • Endcapped for improved performance
  • Low silanol activity
  • Spherical particle shape
  • 5 μm particle size
  • 130 Å pore size
  • Method validation kit format
  • Designed for HPLC systems with CSH particle technology
  • 2.1 mm inner diameter and 150 mm length
  • Units per package: 3