Learn More
Waters Corp XSelect CSH C18 OBD Prep Column, 130Å, 10 μm, 50 mm X 100 mm
Catalog No. 502391724 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect CSH C18 columns utilize Charged Surface Hybrid (CSH) Technology and C18 chemistry for enhanced chromatographic performance. They are suitable for peptide mapping, general peptide separation, purifying target compounds, and are compatible with LC-MS/MS systems.
  • Provides excellent peak shape, high efficiency, and loading capacity for basic compounds.
  • Enables seamless method transfer between HPLC and UPLC™ system platforms.
  • Manufactured in a cGMP, ISO 9001 certified facility.
  • Undergoes rigorous chromatographic testing for reproducible performance.
  • Individually tested for packed bed efficiency; performance chromatogram available.
  • Withstands pressures up to 6000 psi (400 bar).
  • Recommended for operation at temperatures up to 80 °C.
Catalog No. 50-239-1724 Supplier Waters Corp Supplier No. 186007302
Add to Cart
Add to Cart
Request a Quote
XSelect CSH C18 columns utilize Charged Surface Hybrid (CSH) Technology and C18 chemistry for enhanced chromatographic performance. They are suitable for peptide mapping, general peptide separation, purifying target compounds, and are compatible with LC-MS/MS systems.
  • Provides excellent peak shape, high efficiency, and loading capacity for basic compounds.
  • Enables seamless method transfer between HPLC and UPLC™ system platforms.
  • Manufactured in a cGMP, ISO 9001 certified facility.
  • Undergoes rigorous chromatographic testing for reproducible performance.
  • Individually tested for packed bed efficiency; performance chromatogram available.
  • Withstands pressures up to 6000 psi (400 bar).
  • Recommended for operation at temperatures up to 80 °C.