Learn More
Waters Corp Waters XSelect CSH C18 OBD Prep Column, 10 μm, 130 Å, 10 x 150 mm
Catalog No. 502392064 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect CSH C18 OBD Prep Column is designed to maximize productivity when scaling up screening methods to preparative HPLC. It enables direct scale-up from UPLC, UHPLC, or HPLC screening to lab-scale purification.
  • Utilizes Optimum Bed Density (OBD™) Preparative Columns design and packing process for predictable, uniform density profiles, leading to excellent stability, reproducibility, and efficiency.
  • Exhibits exceptional resistance to mechanical chromatographic bed failure and delivers consistent column-to-column performance, extending column lifetimes.
  • CSH Technology provides added selectivity and low pH loadability.
Catalog No. 50-239-2064 Supplier Waters Corp Supplier No. 186008270
The XSelect CSH C18 OBD Prep Column is designed to maximize productivity when scaling up screening methods to preparative HPLC. It enables direct scale-up from UPLC, UHPLC, or HPLC screening to lab-scale purification.
  • Utilizes Optimum Bed Density (OBD™) Preparative Columns design and packing process for predictable, uniform density profiles, leading to excellent stability, reproducibility, and efficiency.
  • Exhibits exceptional resistance to mechanical chromatographic bed failure and delivers consistent column-to-column performance, extending column lifetimes.
  • CSH Technology provides added selectivity and low pH loadability.