Learn More
Waters Corp Waters Corporation XSelect Preparative Column, CSH C18, OBD Column, 10 x 50 mm, 10 μm
Catalog No. 502391621 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
MaxPeak Premier OBD Preparative Columns with MaxPeak High Performance Surfaces enable scientists to purify compounds with confidence by minimizing component/column surface interactions that can lead to target sample loss. Non-specific adsorption (NSA) of metal sensitive compounds is characteristic of certain compound classes such as lipids, organic acids, acidic peptides, oligonucleotides.
  • MaxPeak High Performance Surfaces reduce non-specific adsorption (NSA) and mitigate the loss of metal sensitive components (targets or impurities).
  • Improves peak shape and resolution for better compound detection and fraction collection.
  • Available in different particle sizes and chemistries for the reversed-phase purification of drug compounds, peptides, oligonucleotides, or other small molecules found in materials, biological samples, or natural product sources.
Catalog No. 50-239-1621 Supplier Waters Corp Supplier No. 186008268
Add to Cart
Add to Cart
MaxPeak Premier OBD Preparative Columns with MaxPeak High Performance Surfaces enable scientists to purify compounds with confidence by minimizing component/column surface interactions that can lead to target sample loss. Non-specific adsorption (NSA) of metal sensitive compounds is characteristic of certain compound classes such as lipids, organic acids, acidic peptides, oligonucleotides.
  • MaxPeak High Performance Surfaces reduce non-specific adsorption (NSA) and mitigate the loss of metal sensitive components (targets or impurities).
  • Improves peak shape and resolution for better compound detection and fraction collection.
  • Available in different particle sizes and chemistries for the reversed-phase purification of drug compounds, peptides, oligonucleotides, or other small molecules found in materials, biological samples, or natural product sources.