Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Prep Guard Cartridge, CSH C18, 10 μm, 19 x 10 mm

Catalog No. 502391013 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Waters Prep Guard Cartridge, CSH C18, 10 μm, 19 x 10 mm

Catalog No. 50-239-1013 Supplier Waters Corp Supplier No. 186007290
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Waters Prep Guard Cartridge, CSH C18, 10 μm, 19 x 10 mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.