Learn More
Waters Corp VanGuard Cartridge, C18, 130 Å, 3.5 μm, 2.1 mm X 5 mm
Catalog No. 502391636 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect CSH C18 VanGuard Cartridges extend analytical column lifetime and performance by removing particulate contamination. They are optimized to protect 2.1 mm and 3.0 mm I.D. XSelect CSH C18 analytical columns with 3.5 μm sorbent particles.
  • Extends analytical column lifetime and performance.
  • Removes particulate and chemical contaminants.
  • Protects 2.1 mm, 3.0 mm, and 4.6 mm I.D. analytical columns with 3.5 μm sorbent particles.
  • Maintains UPLC, UHPLC, and HPLC separation.
  • Sustains high pressures in ACQUITY UPLC System applications.
  • Features BEH Technology C18, 130 Å pore size, and 3.5 μm particles.
  • Q.C. tested with peptide map for consistent performance.
  • Quick connection for UPLC column intake.
  • Designed for reversed-phase liquid chromatography.
Catalog No. 50-239-1636 Supplier Waters Corp Supplier No. 186007811
XSelect CSH C18 VanGuard Cartridges extend analytical column lifetime and performance by removing particulate contamination. They are optimized to protect 2.1 mm and 3.0 mm I.D. XSelect CSH C18 analytical columns with 3.5 μm sorbent particles.
  • Extends analytical column lifetime and performance.
  • Removes particulate and chemical contaminants.
  • Protects 2.1 mm, 3.0 mm, and 4.6 mm I.D. analytical columns with 3.5 μm sorbent particles.
  • Maintains UPLC, UHPLC, and HPLC separation.
  • Sustains high pressures in ACQUITY UPLC System applications.
  • Features BEH Technology C18, 130 Å pore size, and 3.5 μm particles.
  • Q.C. tested with peptide map for consistent performance.
  • Quick connection for UPLC column intake.
  • Designed for reversed-phase liquid chromatography.