Learn More
Waters Corp Waters Column, C18, 130Å, 2.5 μm, 3 mm X 50 mm
Catalog No. 50179519 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect CSH C18 columns are general-purpose C18 columns, offering alternative selectivity and superior performance for HPLC, UPLC, and UHPLC applications. Featuring Charged Surface Hybrid (CSH) technology, these 2.5 μm eXtended Performance (XP) columns ensure excellent peak shape and increased loading capacity for basic compounds, along with robust, high-throughput analysis.
  • Provides alternative selectivity to other C18 sorbents.
  • CSH technology ensures superior peak shape and increased loading capacity for basic compounds.
  • XP columns deliver exceptional separation performance, robustness, and high throughput.
  • Fully compatible with all HPLC, UPLC, and UHPLC technologies.
  • Enhances acidic chemical retention and allows maximum flexibility in method creation.
Catalog No. 50-179-519 Supplier Waters Corp Supplier No. 186006105
XSelect CSH C18 columns are general-purpose C18 columns, offering alternative selectivity and superior performance for HPLC, UPLC, and UHPLC applications. Featuring Charged Surface Hybrid (CSH) technology, these 2.5 μm eXtended Performance (XP) columns ensure excellent peak shape and increased loading capacity for basic compounds, along with robust, high-throughput analysis.
  • Provides alternative selectivity to other C18 sorbents.
  • CSH technology ensures superior peak shape and increased loading capacity for basic compounds.
  • XP columns deliver exceptional separation performance, robustness, and high throughput.
  • Fully compatible with all HPLC, UPLC, and UHPLC technologies.
  • Enhances acidic chemical retention and allows maximum flexibility in method creation.