Learn More
Waters Corp XSelect CSH C18 XP Column, 130Å, 2.5 µm, 3 mm X 100 mm, 3/pk
Supplier:  Waters Corp 176002601
Encompass_Preferred
XSelect CSH C18 columns are general purpose C18 columns that provide alternative selectivity compared to other C18 sorbents. Charged Surface Hybrid (CSH) technology enables superior peak shape and increased loading capacity for basic compounds. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
Catalog No. 50-179-260