Learn More
Waters Corp WATERS CORPORATION Column, CSH C18, 2.5 μm, 3 mm × 75 mm
Catalog No. 50179520 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect CSH C18 columns offer alternative selectivity using Charged Surface Hybrid (CSH) technology for superior peak shape and increased loading capacity of basic compounds. These 2.5 μm XP columns, packed in ultra-low dispersion hardware, deliver exceptional separation, robustness, and throughput for HPLC assays, aiding UPLC transition.
  • Provides better resolution and isolation of complex samples.
  • Compatible with HPLC, UPLC, and UHPLC technologies.
  • Ensures consistent and repeatable results.
  • CSH technology addresses peak asymmetry, poor loading, and column bleed.
  • Delivers superior performance with comparable backpressure to core-shell columns.
  • Enhances liquid chromatography in acidic, low ionic strength mobile phases.
  • Offers maximum method development flexibility, especially with low pH mobile phases.
  • Features ultra-low dispersion hardware for superb separation.
Catalog No. 50-179-520 Supplier Waters Corp Supplier No. 186006106
XSelect CSH C18 columns offer alternative selectivity using Charged Surface Hybrid (CSH) technology for superior peak shape and increased loading capacity of basic compounds. These 2.5 μm XP columns, packed in ultra-low dispersion hardware, deliver exceptional separation, robustness, and throughput for HPLC assays, aiding UPLC transition.
  • Provides better resolution and isolation of complex samples.
  • Compatible with HPLC, UPLC, and UHPLC technologies.
  • Ensures consistent and repeatable results.
  • CSH technology addresses peak asymmetry, poor loading, and column bleed.
  • Delivers superior performance with comparable backpressure to core-shell columns.
  • Enhances liquid chromatography in acidic, low ionic strength mobile phases.
  • Offers maximum method development flexibility, especially with low pH mobile phases.
  • Features ultra-low dispersion hardware for superb separation.