Learn More
Waters Corp WATERS CORPORATION Column, C18, 2.5 μm, 4.6 mm, 75 mm
Catalog No. 50201540 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect CSH C18 columns are general-purpose C18 columns offering alternative selectivity and superior peak shape with Charged Surface Hybrid (CSH) technology. These 2.5 μm eXtended Performance (XP) columns are packed in ultra-low dispersion hardware, enabling exceptional separation performance, robustness, and throughput for HPLC assays and UPLC adoption.
  • Offers diverse analyte selectivity to separate closely eluting sample constituents.
  • Overcomes common column-related issues like peak shape asymmetry, poor loading for basic compounds, column bleed, and slow mobile-phase re-equilibration.
  • Provides improved loading capacity and particle size scalability.
  • Non-end-capped sorbent enhances retention of acidic compounds, especially with low pH mobile phases.
  • Withstands 18000 psi (1240 Bar) for 2.5 μm particle size and 130 Å pore size.
Catalog No. 50-201-540 Supplier Waters Corp Supplier No. 186006110
XSelect CSH C18 columns are general-purpose C18 columns offering alternative selectivity and superior peak shape with Charged Surface Hybrid (CSH) technology. These 2.5 μm eXtended Performance (XP) columns are packed in ultra-low dispersion hardware, enabling exceptional separation performance, robustness, and throughput for HPLC assays and UPLC adoption.
  • Offers diverse analyte selectivity to separate closely eluting sample constituents.
  • Overcomes common column-related issues like peak shape asymmetry, poor loading for basic compounds, column bleed, and slow mobile-phase re-equilibration.
  • Provides improved loading capacity and particle size scalability.
  • Non-end-capped sorbent enhances retention of acidic compounds, especially with low pH mobile phases.
  • Withstands 18000 psi (1240 Bar) for 2.5 μm particle size and 130 Å pore size.