Learn More
Waters Corp Waters XSelect CSH C18 XP Column, C18, 2.5 μm, 130 Å, 2.1 mm × 50 mm
Supplier:  Waters Corp 186006101
Encompass_Preferred
XSelect CSH C18 XP Columns utilize Charged Surface Hybrid (CSH) technology to offer alternative selectivity and superior peak shape for basic compounds. These 2.5 μm columns deliver exceptional separation performance for HPLC assays, facilitating transition to UPLC and UHPLC technologies, and enabling higher resolution for complex samples.
  • Offers alternative selectivity for C18 separations.
  • Charged Surface Hybrid (CSH) technology provides superior peak shape for basic compounds.
  • Increased loading capacity for basic compounds.
  • Exceptional separation performance and throughput for HPLC assays.
  • Seamless transition path to UPLC adoption.
  • Fully compatible with HPLC, UPLC, and UHPLC technologies.
  • Excellent batch-to-batch reproducibility.
Catalog No. 50-179-515