Learn More
Waters Corp Waters Column, CSH C18, 130Å, 2.5 μm
Supplier: Waters Corp 176002596
XSelect CSH C18 columns are general-purpose C18 columns that offer alternative selectivity compared to other C18 sorbents. They incorporate Charged Surface Hybrid (CSH) technology, which improves peak shape and increases loading capacity for basic compounds. These 2.5 μm eXtended Performance (XP) columns are packed in ultra-low dispersion hardware, delivering exceptional separation performance, robustness, and throughput for HPLC assays, while also providing a smooth transition path to UPLC adoption.
- Chemistry: C18
- Particle technology: CSH
- Particle size: 2.5 μm
- Pore size: 130 Å
- Inner diameter: 2.1 mm
- Length: 75 mm
- Maximum pressure: 18000 psi (1240 Bar)
- System compatibility: UPLC, UHPLC
- Units per package: 3
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.