Learn More
Waters Corp Flangeless Cartridge, Oasis HLB, 1 cc/10 mg, 30 μm
Supplier:  Waters Corp 186006237
Encompass_Preferred
The Oasis HLB Flangeless Cartridge is a solid-phase extraction product featuring a hydrophilic-lipophilic balanced reversed-phase sorbent. It is designed for sample preparation in various laboratory applications.
  • Hydrophilic-lipophilic balanced reversed-phase sorbent
  • Dimensions: 1 cc/10 mg
  • Particle size: 30 μm
  • Supplied in a box of 100 cartridges
Catalog No. 50-201-588