Learn More
Waters Corp Waters XSelect CSH C18 XP Column, 130Å, 2.5 μm, 4.6 mm × 100 mm
Supplier:  Waters Corp 186006111
Encompass_Preferred
The XSelect CSH C18 XP Column is a general-purpose C18 column with Charged Surface Hybrid (CSH) technology, offering alternative selectivity and superior peak shape for basic compounds. These 2.5 μm XP columns provide exceptional separation performance, robustness, and throughput for HPLC assays, supporting transition to UPLC.
  • Diverse analyte selectivity for closely eluting constituents.
  • Suitable for high-resolution separation of complex samples.
  • Enhances resolution, saving time in assay development.
  • Compatible with HPLC, UPLC, and UHPLC technologies.
  • Outstanding performance, equivalent backpressure to core-shell columns.
  • Improved loading capacity and particle size scalability with CSH technology.
  • CSH technology enhances performance in acidic, low ionic strength mobile phases.
Catalog No. 50-201-541