Learn More
Waters Corp Waters Column, XSelect CSH C18, 130Å, 2.5 μm, 2.1 mm x 30 mm
Catalog No. 50179253 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect CSH C18 columns are general-purpose C18 columns with Charged Surface Hybrid (CSH) technology, offering superior peak shape and increased loading capacity for basic compounds. These 2.5 μm eXtended Performance (XP) columns provide exceptional separation performance, robustness, and throughput for HPLC assays, and enable a seamless transition to UPLC.
  • C18 chemistry with reversed phase separation.
  • Hybrid particle substrate.
  • pH range of 1 to 11.
  • Maximum pressure of 18000 psi (1240 Bar).
  • Spherical particles with 2.5 μm size.
  • Pore size of 130 Å.
  • Inner diameter of 2.1 mm and length of 30 mm.
  • Compatible with UPLC and UHPLC systems.
  • Includes eCord.
  • Supplied as 3 units per package.
Catalog No. 50-179-253 Supplier Waters Corp Supplier No. 176002594
XSelect CSH C18 columns are general-purpose C18 columns with Charged Surface Hybrid (CSH) technology, offering superior peak shape and increased loading capacity for basic compounds. These 2.5 μm eXtended Performance (XP) columns provide exceptional separation performance, robustness, and throughput for HPLC assays, and enable a seamless transition to UPLC.
  • C18 chemistry with reversed phase separation.
  • Hybrid particle substrate.
  • pH range of 1 to 11.
  • Maximum pressure of 18000 psi (1240 Bar).
  • Spherical particles with 2.5 μm size.
  • Pore size of 130 Å.
  • Inner diameter of 2.1 mm and length of 30 mm.
  • Compatible with UPLC and UHPLC systems.
  • Includes eCord.
  • Supplied as 3 units per package.