Learn More
Waters Corp Waters Corporation XSelect CSH C18 XP Column, 2.5 μm, 130 Å, 2.1 x 30 mm
Catalog No. 50179514 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These general-purpose C18 columns use Charged Surface Hybrid (CSH) technology for superior peak shape and increased loading capacity of basic compounds. Packed in ultra-low dispersion hardware, 2.5 μm XP columns provide exceptional separation, robustness, and throughput for HPLC assays. Compatible with HPLC, UPLC, and UHPLC, they optimize total cycle time and improve analysis productivity.
  • Offers alternative selectivity.
  • Charged Surface Hybrid (CSH) technology for superior peak shape and increased loading capacity.
  • Ultra-low dispersion hardware.
  • 2.5 μm XP columns deliver exceptional separation and throughput.
  • Seamless transition to UPLC.
  • Optimizes total cycle time without compromising resolution.
  • Compatible with HPLC, UPLC, and UHPLC.
  • Improves HPLC analysis productivity by up to 4 times.
  • 2.1 mm I.D variants withstand pressures up to 18,000 psi.
Catalog No. 50-179-514 Supplier Waters Corp Supplier No. 186006100
These general-purpose C18 columns use Charged Surface Hybrid (CSH) technology for superior peak shape and increased loading capacity of basic compounds. Packed in ultra-low dispersion hardware, 2.5 μm XP columns provide exceptional separation, robustness, and throughput for HPLC assays. Compatible with HPLC, UPLC, and UHPLC, they optimize total cycle time and improve analysis productivity.
  • Offers alternative selectivity.
  • Charged Surface Hybrid (CSH) technology for superior peak shape and increased loading capacity.
  • Ultra-low dispersion hardware.
  • 2.5 μm XP columns deliver exceptional separation and throughput.
  • Seamless transition to UPLC.
  • Optimizes total cycle time without compromising resolution.
  • Compatible with HPLC, UPLC, and UHPLC.
  • Improves HPLC analysis productivity by up to 4 times.
  • 2.1 mm I.D variants withstand pressures up to 18,000 psi.